missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Annexin V-Biotin Apoptosis Detection Kit

Annexin V staining, paired with 7-AAD or PI is widely used to identify apoptotic stages by flow cytometry.

$145.20 - $508.20

Specifications

For Use With (Application) Flow Cytometric Analysis
Synonym Lipocortin V, Endonexin II, Calphobindin I
Assay > 98%
Conjugate Biotin
Formula Weight 35.8 kd
View More Specs

Disclaimers:

For Research Use Only


Includes: Annexin V-Biotin protein
Binding Buffer (4x)
Propidium Iodide (20 ug/mL)


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509295 Life Technologies
LSBMS500BT20
20 tests Each for $145.20
509294 Life Technologies
LSBMS500BT100
100 tests Each for $302.28
509296 Life Technologies
LSBMS500BT300
300 tests Each for $508.20
Specifications Description & Specifications

Specifications

Flow Cytometric Analysis
> 98%
35.8 kd
Lipocortin V, Endonexin II, Calphobindin I
Biotin