missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Annexin V Apoptosis Detection Set PE-Cyanine7

Due to the emission spectrum of PE-Cyanine7, not compatible with propidium iodide and 7-AAD. It is recommended to substitute a Fixable Viability Dye such as eFluor™ 660 or eFluor™ 780 in their place.

$265.35 - $590.15

Specifications

Reactivity Human, Mouse, Rat
Storage Buffer aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Storage Requirements 2° to 8°C. Do not freeze. Light sensitive material.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50246345 Life Technologies
LS88810372
50 tests Each for $265.35
50246346 Life Technologies
LS88810374
200 tests Each for $590.15
Specifications

Description

Can be used at 5μL/test
Description & Specifications

Specifications

Human, Mouse, Rat
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
2° to 8°C. Do not freeze. Light sensitive material.