missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Annexin V Apoptosis Detection Kit PerCP-eFluor™ 710

Pre-titrated and tested on mouse thymocytes cultured overnight in medium (to induce apoptosis) or on Jurkat cells treated with 10 μM Camptothecin for 4 hours. This can be used at 5μL per test.

$262.68 - $471.24

Specifications

For Use With (Application) Flow Cytometric Analysis
Storage Buffer Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Storage Requirements 2° to 8°C. Do not freeze. Light sensitive material.
View More Specs

Alert:

Contains Sodium azide

Disclaimers:

For Research Use Only


Includes: Cat. No. 88-8008-72:
10X Binding Buffer
Annexin V PerCP-eFluor™ 710 : 5μL/test, 50 tests Protect from light.
Cat. No. 88-8008-74:
10X Binding Buffer
Annexin V PerCP-eFluor™ 710 : 5μL/test, 200 tests Protect from light.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509280 Life Technologies
LS88800872
50 tests Each for $262.68
509281 Life Technologies
LS88800874
200 tests Each for $471.24
Specifications

Description

Fixable Viability Dye eFluor™ 450 is not recommended for use with Annexin V Apoptosis Detection Kits.
Description & Specifications

Specifications

Flow Cytometric Analysis
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
2° to 8°C. Do not freeze. Light sensitive material.