missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Annexin V Apoptosis Detection Kit FITC

Compatible with intracellular staining. Pre-titrated and tested on mouse thymocytes cultured overnight in medium (to induce apoptosis) or on Jurkat cells treated with 10 μM Camptothecin for 4 hours. Both the Annexin V FITC and the Propidium Iodide components can be used at 5μL per test

$174.24 - $531.96

Specifications

For Use With (Application) Flow Cytometric Analysis
Storage Buffer 50mM Tris, pH 7.4, 100mM NaCl, 1% BSA, 0.02% Sodium Azide
Storage Requirements 2° to 8°C. Do not freeze. Light sensitive material.
View More Specs

Alert:

Contains Sodium azide

Disclaimers:

For Research Use Only


Includes: Cat. No. 88-8005-72:
10X Binding Buffer
Annexin V FITC : 5μL/test, 50 tests Protect from light.
Propidium Iodide Staining Solution : 5μL/test, 100 tests Protect from light.
Cat. No. 88-8005-74:
10X Binding Buffer
Annexin V FITC : 5μL/test, 200 tests Protect from light.
Propidium Iodide Staining Solution : 5μL/test, 2x100 tests Protect from light.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509275 Life Technologies
LS88800574
200 tests Each for $531.96
501128958 Life Technologies
LS88800572
50 tests Each for $174.24
Specifications

Description

Fixable Viability Dye eFluor™ 450 is not recommended for use with Annexin V Apoptosis Detection Kits.
Description & Specifications

Specifications

Flow Cytometric Analysis
50mM Tris, pH 7.4, 100mM NaCl, 1% BSA, 0.02% Sodium Azide
2° to 8°C. Do not freeze. Light sensitive material.