missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

eBioscience™ Annexin V Apoptosis Detection Kit eFluor™ 450

Compatible with intracellular staining. Pre-titrated and tested on mouse thymocytes cultured overnight in medium (to induce apoptosis) or on Jurkat cells treated with 10 μM Camptothecin for 4 hours. Both the Annexin V eFluor 450 and the 7-AAD components can be used at 5μL per test.

$241.56 - $429.00

Specifications

For Use With (Application) Flow Cytometric Analysis
Storage Buffer Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Storage Requirements 2° to 8°C. Do not freeze. Light sensitive material.
View More Specs

Alert:

Contains Sodium azide

Disclaimers:

For Research Use Only


Includes: Cat. No. 88-8006-72:
10X Binding Buffer
Annexin V eFluor™ 450 : 5μL/test, 50 tests Protect from light.
7-AAD Viability Staining Solution : 5μL/test, 100 tests Protect from light.
Cat. No. 88-8006-74:
10X Binding Buffer
Annexin V eFluor™ 450 : 5μL/test, 200 tests Protect from light.
7-AAD Viability Staining Solution : 5μL/test, 2x100 tests Protect from light.


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509276 Life Technologies
LS88800672
50 tests Each for $241.56
509277 Life Technologies
LS88800674
200 tests Each for $429.00
Specifications Description & Specifications

Specifications

Flow Cytometric Analysis
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
2° to 8°C. Do not freeze. Light sensitive material.