Please login to your online account to display your discounted pricing

eBioscience Annexin V Apoptosis Detection Kit APC

Compatible with intracellular staining. Pre-titrated and tested on mouse thymocytes cultured overnight in medium (to induce apoptosis) or on Jurkat cells treated with 10 μM Camptothecin for 4 hours. Both the Annexin V APC and the Propidium iodide components can be used at 5μL per test

$216.05 - $638.00

Specifications

For Use With (Application) Flow Cytometric Analysis
Storage Buffer Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Storage Requirements 2° to 8°C. Do not freeze. Light sensitive material.
View More Specs
Includes:  Cat. No. 88-8007-72:
10X Binding Buffer
Annexin V APC : 5μL/test, 50 tests Protect from light.
Propidium Iodide Staining Solution : 5μL/test, 100 tests Protect from light.
Cat. No. 88-8007-74:
10X Binding Buffer
Annexin V APC : 5μL/test, 200 tests Protect from light.
Propidium Iodide Staining Solution : 5μL/test, 2x100 tests Protect from light.

 Alert

Contains Sodium azide

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509279

 
ebioscience
88800774
200 tests Each for $638.00

501129048

 
ebioscience
88800772
50 tests Each for $216.05
Description & Specifications

Specifications

For Use With (Application) Flow Cytometric Analysis
Storage Buffer Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Storage Requirements 2° to 8°C. Do not freeze. Light sensitive material.

Fixable Viability Dye eFluor™ 450 is not recommended for use with Annexin V Apoptosis Detection Kits.