missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Annexin 1, Certified Mass Spec grade

For research use

Specifications

Formulation Lyophilized powder
Recombinant No
Storage Requirements -30° to -10°C
Product Type Annexin 1, Certified Mass Spec grade
Format Lyophilized Powder
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67732ODL Alfa Aesar™
J67732ODL
3 vials N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Specifications

Lyophilized powder
-30° to -10°C
Lyophilized Powder
No
Annexin 1, Certified Mass Spec grade
RUO