Please login to your online account to display your discounted pricing

Alfa Aesar™ Annexin 1, Certified Mass Spec grade


For research use

Manufacturer: alfa aesar™  J67732LPL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67732LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1 vial
Recombinant No
Storage Requirements -30° to -10°C
Regulatory Status RUO
Protein Length AMVSEFLKQAW
Product Type Annexin 1, Certified Mass Spec grade
Format Lyophilized powder
Formulation Lyophilized powder