Please login to your online account to display your discounted pricing

Anisomycin, Tocris

Activates JNK/SAPK/p38 MAP kinase

$116.01 - $451.04

Chemical Identifiers

Chemical Name or Material Anisomycin
Assay Percent Range >98%
Molecular Formula C14H19NO4
Formula Weight 265.31
Synonym (2R,3S,4S)-2-[(4-Methoxyphenyl)methyl]-3,4-pyrrolidinediol 3-acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

129010

 
r & d systems
129010
10mg Each for $116.01

129050

 
r & d systems
129050
50mg Each for $451.04
Description & Specifications

Specifications

Formula Weight 265.31
Quantity 10mg
Molecular Formula C14H19NO4
Chemical Name or Material Anisomycin
Assay Percent Range >98%
CAS 22862-76-6
Synonym (2R,3S,4S)-2-[(4-Methoxyphenyl)methyl]-3,4-pyrrolidinediol 3-acetate
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C