missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Anisomycin, 97%

$380.91 - $380.91

Chemical Identifiers

CAS 22862-76-6
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD00077650
InChI Key YKJYKKNCCRKFSL-RDBSUJKOSA-N
Synonym (2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)-3-pyrrolidinyl acetate; Flagecidin
PubChem CID 253602
ChEBI CHEBI:338412
IUPAC Name [(2R,3S,4S)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl] acetate
SMILES CC(=O)OC1C(CNC1CC2=CC=C(C=C2)OC)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62964MF View Documents Alfa Aesar™
J62964MF
50mg Each for $380.91
 
Description

Description

  • A protein synthesis inhibitor
  • Causes apoptosis of PC12 Cells
Specifications

Chemical Identifiers

22862-76-6
265.309
YKJYKKNCCRKFSL-RDBSUJKOSA-N
253602
[(2R,3S,4S)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl] acetate
C14H19NO4
MFCD00077650
(2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)-3-pyrrolidinyl acetate; Flagecidin
CHEBI:338412
CC(=O)OC1C(CNC1CC2=CC=C(C=C2)OC)O

Specifications

22862-76-6
MFCD00077650
20705
(2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)-3-pyrrolidinyl acetate; Flagecidin
YKJYKKNCCRKFSL-RDBSUJKOSA-N
[(2R,3S,4S)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl] acetate
253602
265.31
97%
140°C to 141°C
Anisomycin
C14H19NO4
UN3462
14,670
Soluble in water at 2 mg/ml, in DMSO at 20mg/ml, and in methanol at 20mg/ml
CC(=O)OC1C(CNC1CC2=CC=C(C=C2)OC)O
265.309
CHEBI:338412
Powder
White
50mg
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.

RTECSNumber : BZ9800000

Recommended Storage : −30° to −10°C