Please login to your online account to display your discounted pricing

Alfa Aesar™ Anisomycin, 10mg/mL in DMSO, sterile-filtered

Chemical Identifiers

Chemical Name or Material Anisomycin
CAS 22862-76-6
Name Note 10mg/mL in DMSO, sterile-filtered
MDL Number MFCD00077650
Synonym Flagecidin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67498XF

 
alfa aesar™
J67498XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Anisomycin
CAS 22862-76-6
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note 10mg/mL in DMSO, sterile-filtered
MDL Number MFCD00077650
Synonym Flagecidin
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
RTECS Number BZ9800000
Physical Form Liquid
Storage Note 1 -30° to -10°C
Quantity 1mL

Applications
It is applied as an antiparasitic agent and antineoplastic agent. It acts as a protein synthesis inhibitor (blocks translation), potent activator of stress-activated protein kinases (JNK/SAPK) and p38 MAP kinase. It also scts as a potent signaling agonist to selectively elicit homologous desensitization of immediate early gene induction (c-fos, fosB, c-jun, junB and junD). Activates mitogen-activated protein (MAP) kinases (JNK/SAPK and p38/RK).

Solubility
Fully miscible in water. Soluble to 50 mM in ethanol and to 100 mM in DMSO. Soluble in methanol (20 mg/ml clear, faint yellow solution).Slightly soluble in alcohols, esters, ketones, chloroform, benzene, toluene, hexane, and ethyl acetate (10 mg/ml clear, colorless solution), DMF (20 mg/ml), and PBS pH 7.2 (0.5 mg/ml).

Notes
Protect from heat, acids, reducing agents, acid chlorides and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated at -20°C and keep ignition sources away.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.

-30° to -10°C