missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Anisole, 99%, pure, ACROS Organics™

Manufacturer:  Acros Organics 153922500

 View more versions of this product

Catalog No. AC153922500



Specifications

Specifications

Anisole
43°C
Glass bottle
0.99
154.0°C
-37.0°C
100-66-3
C7H8O
methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene, benzene, methoxy
RDOXTESZEPMUJZ-UHFFFAOYSA-N
anisole
7519
108.14
99%
0.9900g/mL
Authentic
1.5160 to 1.5190
0.1% Max. (K.F.)
Colorless
250mL
99%
C6H5OCH3
Solubility in water: 1.6g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone
COC1=CC=CC=C1
108.14
CHEBI:16579
Liquid
Pure

Chemical Identifiers

100-66-3
108.14
methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene, benzene, methoxy
CHEBI:16579
COC1=CC=CC=C1
C7H8O
RDOXTESZEPMUJZ-UHFFFAOYSA-N
7519
anisole
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour. Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Store in a well-ventilated place. Keep container tightly closed.

Warning

SDS Documents