missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Anisole, 99%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 153920010

 View more versions of this product

Catalog No. AC153920010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Anisole
-37.0°C
0.9900g/mL
43°C
98.5% min. (GC)
C7H8O
C6H5OCH3
06,138
0.99
methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
RDOXTESZEPMUJZ-UHFFFAOYSA-N
anisole
7519
108.14
Pure
100-66-3
Colorless
154.0°C
Authentic
Glass bottle
1.5160 to 1.5190
1 L
01,41
15,662
Solubility in water: 1.6g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone
COC1=CC=CC=C1
108.14
CHEBI:16579
99%
Liquid

Chemical Identifiers

100-66-3
108.14
methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
CHEBI:16579
COC1=CC=CC=C1
C7H8O
RDOXTESZEPMUJZ-UHFFFAOYSA-N
7519
anisole
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ven

GHS Signal Word: Warning

EINECSNumber : 202-876-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only