missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Anisil, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1059406
Description
Anisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAnisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Anisil | |
132°C to 134°C | |
MFCD00008405 | |
531345 | |
Insoluble in water. | |
COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC | |
270.284 | |
270.29 |
1226-42-2 | |
C16H14O4 | |
5 g | |
4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione | |
YNANGXWUZWWFKX-UHFFFAOYSA-N | |
1,2-bis(4-methoxyphenyl)ethane-1,2-dione | |
71043 | |
≥98% |
Chemical Identifiers
1226-42-2 | |
270.284 | |
YNANGXWUZWWFKX-UHFFFAOYSA-N | |
71043 | |
COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC |
C16H14O4 | |
MFCD00008405 | |
4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione | |
1,2-bis(4-methoxyphenyl)ethane-1,2-dione |
Safety and Handling
EINECSNumber : 214-960-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only