Please login to your online account to display your discounted pricing

Aniracetam, Tocris

Desensitization inhibitor (AMPA > kainate)

$164.61 - $649.50

Chemical Identifiers

Chemical Name or Material Aniracetam
Assay Percent Range >99%
Molecular Formula C12H13NO3
Formula Weight 219.24
Synonym 1-(4-Methoxybenzoyl)-2-pyrrolidinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

0867250

 
r & d systems
RDS0867250
50mg Each for $649.50

086750

 
r & d systems
RDS086750
50mg Each for $164.61
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 219.24
Chemical Name or Material Aniracetam
Assay Percent Range >99%
Molecular Formula C12H13NO3
CAS 72432-10-1
Synonym 1-(4-Methoxybenzoyl)-2-pyrrolidinone
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature