Please login to your online account to display your discounted pricing

Aniracetam, Tocris


Desensitization inhibitor (AMPA > kainate)

Quantity 50mg

Manufacturer: r & d systems  RDS086750

 View more versions of this product

Catalog No. 086750

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 219.24
Chemical Name or Material Aniracetam
Assay Percent Range >99%
Molecular Formula C12H13NO3
CAS 72432-10-1
Synonym 1-(4-Methoxybenzoyl)-2-pyrrolidinone
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Aniracetam
Assay Percent Range >99%
Molecular Formula C12H13NO3
Formula Weight 219.24
Synonym 1-(4-Methoxybenzoyl)-2-pyrrolidinone
Accessories

Store at Room Temperature