Please login to your online account to display your discounted pricing

Alfa Aesar™ Anionic Acid Labile Surfactant II (AALS II), Certified Mass Spec Grade

Chemical Identifiers

Chemical Name or Material Anionic Acid Labile Surfactant II (AALS II)
Purity Grade Certified Mass Spec Grade
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67832LB0

  SDS
alfa aesar™
J67832LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Anionic Acid Labile Surfactant II (AALS II)
Physical Form Lyophilized Powder
Purity Grade Certified Mass Spec Grade
Storage Note 1 -30° to -10°C
Color White
Odor Odorless
Quantity 5mg
Safety and Handling

-30° to -10°C