Please login to your online account to display your discounted pricing

Alfa Aesar™ Anionic Acid Labile Surfactant I (AALS I), Certified Mass Spec Grade


Quantity 5mg

Manufacturer: alfa aesar™  J67647LB0

 View more versions of this product

Catalog No. AAJ67647LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Anionic Acid Labile Surfactant I (AALS I)
Purity Grade Certified Mass Spec Grade
Physical Form Lyophilized Powder
Storage Note 1 -30° to -10°C
Color White
Odor Odorless
Quantity 5mg

Specifications

Chemical Name or Material Anionic Acid Labile Surfactant I (AALS I)
Purity Grade Certified Mass Spec Grade

  • This detergent is effective at solubilizing peptides and proteins, optimizing enzymatic digestions, and reducing surface adsorption losses via non-specific interactions

Accessories

-30° to -10°C

SDS & Safety Documents