Please login to your online account to display your discounted pricing

Alfa Aesar™ Anionic Acid Labile Surfactant I (AALS I), Certified Mass Spec Grade

Chemical Identifiers

Chemical Name or Material Anionic Acid Labile Surfactant I (AALS I)
Purity Grade Certified Mass Spec Grade
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67647LB0

 
alfa aesar™
J67647LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Anionic Acid Labile Surfactant I (AALS I)
Purity Grade Certified Mass Spec Grade
Physical Form Lyophilized Powder
Storage Note 1 -30° to -10°C
Color White
Odor Odorless
Quantity 5mg

  • This detergent is effective at solubilizing peptides and proteins, optimizing enzymatic digestions, and reducing surface adsorption losses via non-specific interactions

Safety and Handling

-30° to -10°C