Please login to your online account to display your discounted pricing

Alfa Aesar™ Anilinoacetaldehyde diethyl acetal, 98%

$23.92 - $329.57

Chemical Identifiers

Chemical Name or Material Anilinoacetaldehyde diethyl acetal
CAS 22758-34-5
Molecular Formula C6H5NHCH2CH(OCH2CH3)2
Formula Weight 209.29g/mol
MDL Number MFCD00051601
Beilstein 2806325
Synonym N-Phenylaminoacetaldehyde diethyl acetal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1158703

 
alfa aesar™
A11587-03
1g Each for $23.92

AAA1158714

 
alfa aesar™
A11587-14
25g Each for $329.57
Description & Specifications

Specifications

Boiling Point 153° to 154°C (12mmHg)
Quantity 1g
Chemical Name or Material Anilinoacetaldehyde diethyl acetal
CAS 22758-34-5
Assay 98%
Molecular Formula C6H5NHCH2CH(OCH2CH3)2
Formula Weight 209.29g/mol
MDL Number MFCD00051601
Beilstein 2806325
Synonym N-Phenylaminoacetaldehyde diethyl acetal
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.