missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aniline sulfate, 98%

Manufacturer:  Alfa Aesar™ A155630E

 View more versions of this product

Catalog No. AAA155630E



Specifications

Specifications

Aniline sulfate
2500g
(C6H5NH2)2·H2SO4
UN2811
Benzenamine sulfate 2:1
284.34
Odorless
542-16-5
MFCD00013110
3729533
Soluble in water
98%

Chemical Identifiers

542-16-5
(C6H5NH2)2·H2SO4
MFCD00013110
Benzenamine sulfate 2:1
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H372-H341-H351-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of causing cancer.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P280-P273-P305+P351+P338-P361-P304+P340-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

Documents