Please login to your online account to display your discounted pricing

Aniline, ACS, 99+%, Alfa Aesar™

$137.26 - $137.26

Chemical Identifiers

Chemical Name or Material Aniline
CAS 62-53-3
Purity Grade ACS Grade
Molecular Formula C6H7N
Formula Weight 93.13
MDL Number MFCD00007629
UN Number UN1547
Beilstein 605631
Merck Index 14,659
Synonym Benzenamine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA36238K2

  SDS
alfa aesar
36238K2
Each for $137.26
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 93.13
Quantity 1L
Assay ≥99%
Chemical Name or Material Aniline
TSCA TSCA
Density 1.021
Physical Form Liquid
Molecular Formula C6H7N
MDL Number MFCD00007629
CAS 62-53-3
UN Number UN1547
Beilstein 605631
Merck Index 14,659
Refractive Index 1.5860
Synonym Benzenamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H372-H341-H351-H318-H227-H317;
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of causing cancer.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.
Boiling Point 183° to 184°C
Melting Point −6°C
Health Hazard 3 GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 70°C (158°F)
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ANILINE
RTECS Number BW6650000
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H372-H341-H351-H318-H227-H317;
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of causing cancer.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.