Learn More
Aniline, 99+%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423420050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aniline | |
-6.2°C | |
1.0213g/mL | |
76°C | |
Authentic | |
Glass bottle | |
1.5845 to 1.5865 | |
MFCD00007629 | |
12, 59 | |
1.0213 | |
benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam | |
PAYRUJLWNCNPSJ-UHFFFAOYSA-N | |
aniline | |
6115 | |
CHEBI:17296 | |
0.5mmHg at 20°C | |
ACS Reagent |
62-53-3 | |
Colorless to White | |
181.0°C to 185.0°C | |
0.005 % max. | |
99% min. (GC) | |
C6H7N | |
C6H5NH2 | |
5 mL | |
06,21; 17,16 | |
15, 652 | |
Solubility in water: 36g/L (20°C). | |
NC1=CC=CC=C1 | |
93.13 | |
4.4 mPa.s (20°C) | |
93.13 | |
99+% | |
Crystalline Powder or Crystals |
Chemical Identifiers
62-53-3 | |
93.13 | |
PAYRUJLWNCNPSJ-UHFFFAOYSA-N | |
6115 | |
aniline |
C6H7N | |
MFCD00007629 | |
benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam | |
CHEBI:17296 | |
NC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Toxic in contact with skin.
Toxic if swallowed.
Causes serious eye dam
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Danger
EINECSNumber : 200-539-3
Recommended Storage : this product darkens to reddish-brown on storage
RUO – Research Use Only