missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aniline, 99.8%, pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC158191000

 View more versions of this product

Catalog No. AC158191000



Specifications

Specifications

Aniline
76°C
Glass bottle
1.0213
4.4 mPa.s (20°C)
Pink to White
100mL
99.75% min. (GC)
C6H5NH2
12, 59
15, 652
Solubility in water: 36g/L (20°C).
C1=CC=C(C=C1)N
93.13
CHEBI:17296
Crystalline Powder, Crystals or Crystalline Mass
Pure
1.0213g/mL
Authentic
1.5845 to 1.5865
0.5mmHg at 20°C
181.0°C to 185.0°C
-6.2°C
62-53-3
C6H7N
MFCD00007629
06,21; 17,16
aminobenzene
PAYRUJLWNCNPSJ-UHFFFAOYSA-N
aniline
6115
93.13
99.8%

Chemical Identifiers

62-53-3
93.13
PAYRUJLWNCNPSJ-UHFFFAOYSA-N
6115
aniline
C6H7N
MFCD00007629
aminobenzene
CHEBI:17296
C1=CC=C(C=C1)N
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Toxic in contact with skin.
Toxic if swallowed.
Causes serious eye dam

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Danger

EINECSNumber : 200-539-3

Recommended Storage : Darkens to reddish-brown on storage

SDS Documents