Learn More
Aniline, 99.5%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 221732500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aniline | |
-6.2°C | |
1.0213g/mL | |
76°C | |
99.4% min. (GC) | |
C6H7N | |
C6H5NH2 | |
250 mL | |
06,21; 17,16 | |
15,652 | |
Solubility in water: 36g/L (20°C). | |
NC1=CC=CC=C1 | |
93.13 | |
4.4 mPa.s (20°C) | |
93.13 | |
Extra Pure |
62-53-3 | |
Brown to Colorless | |
181.0°C to 185.0°C | |
Authentic | |
Glass bottle | |
1.5845 to 1.5865 | |
MFCD00007629 | |
12,59 | |
1.0213 | |
benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam | |
PAYRUJLWNCNPSJ-UHFFFAOYSA-N | |
aniline | |
6115 | |
CHEBI:17296 | |
99.5% | |
Liquid |
Chemical Identifiers
62-53-3 | |
93.13 | |
PAYRUJLWNCNPSJ-UHFFFAOYSA-N | |
6115 | |
aniline |
C6H7N | |
MFCD00007629 | |
benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam | |
CHEBI:17296 | |
NC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Toxic in contact with skin.
Toxic if swallowed.
Causes serious eye dam
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Danger
EINECSNumber : 200-539-3
RUO – Research Use Only