Please login to your online account to display your discounted pricing

Alfa Aesar™ Anhydrotetracycline hydrochloride


Quantity 10mg

Manufacturer: alfa aesar™  J66688MA

 View more versions of this product

Catalog No. AAJ66688MA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 462.88
Melting Point 220° to 223°C
Chemical Name or Material Anhydrotetracycline hydrochloride
MDL Number MFCD00151453
CAS 13803-65-1
Physical Form Crystalline Solid
Beilstein 4228856
Solubility Information Soluble in ethanol, DMSO, or DMF
Storage Note 1 -30° to -10°C
Quantity 10mg
RTECS Number QI7860000
Color Yellow
Sensitivity Moisture Sensitive

Specifications

Chemical Name or Material Anhydrotetracycline hydrochloride
CAS 13803-65-1
Formula Weight 462.88
MDL Number MFCD00151453
Beilstein 4228856

Applications
Anhydrotetracycline hydrochloride acts as a useful effector of tetracycline-dependent gene expression in bacterial expression systems.

Solubility
Soluble in ethanol, DMSO, or DMF

Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Accessories

-30° to -10°C