Please login to your online account to display your discounted pricing

Alfa Aesar™ Angiotensinogen (1-13), Human

For cellular and molecular biology applications

Specifications

Product Type Angiotensinogen (1-13), human
Cross Reactivity Human
Molecular Weight 1645.93
Storage Requirements -30° to -10°C
Sequence Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Val-Ile-His
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66542LB0

 
alfa aesar™
J66542LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66542MCR

 
alfa aesar™
J66542MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Angiotensinogen (1-13), human
Cross Reactivity Human
Molecular Weight 1645.93
Storage Requirements -30° to -10°C
Sequence Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Val-Ile-His
Format Solid
Regulatory Status RUO
Purification Method HPLC