Please login to your online account to display your discounted pricing

Alfa Aesar™ Angiotensinogen (1-13), Human


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66542LB0

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66542LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 5mg
Format Solid
Sequence Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Val-Ile-His
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Angiotensinogen (1-13), human
Molecular Weight 1645.93
Purification Method HPLC
Cross Reactivity Human