Please login to your online account to display your discounted pricing

Alfa Aesar™ Angiotensin I, Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67640LPL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67640LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Product Type Angiotensin I, Certified Mass Spec Grade
Regulatory Status RUO
Format Lyophilized powder
Sequence DRVYIHPFHL
Molecular Weight 1296.40
Quantity 1 vial, 500 pmol
Purification Method HPLC