missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Angiotensin Converting Enzyme-2, His·Tag™, Human, Recombinant, NSO Cells
Description
- Recombinant, human ACE-2 ectodomain (amino acids 1-740) fused at the C-terminus to a His•Tag™ sequence and expressed in NSO cells
- Native ACE-2 is an integral membrane zinc metalloprotease that can be inhibited by EDTA.
- ACE-2 cleaves angiotensins I and II
- By SDS-PAGE, the apparent molecular mass of the glycosylated protein is ∽120 kDa under either reducing or non-reducing condition
Specifications
Specifications
| Concentration | Concentration Label: Please refer to vial label for lot-specific concentration |
| Purity | ≥90% by SDS-PAGE |
| Content And Storage | ≤-70°C |
| Form | Liquid |
| Quantity | 10 μg |
| Synonym | ACE Homolog, ACEH, ACE-2 |
| Product Type | Angiotensin Converting Enzyme (ACE) |