Please login to your online account to display your discounted pricing

Angiogenin Mouse anti-Human, Biotin, Clone: 26-2F, eBioscience™

Mouse Monoclonal Antibody

$131.95 - $332.05

Specifications

Antigen Angiogenin
Clone 26-2F
Host Species Mouse
Gene Alias Ribonuclease 5
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122602

 
life technologies
LS13976280
25μg Each for $131.95

501122603

 
life technologies
LS13976282
100μg Each for $332.05
Description & Specifications

Specifications

Antigen Angiogenin
Clone 26-2F
Host Species Mouse
Gene Alias Ribonuclease 5
Species Reactivity Human
Applications Enzyme Linked Immunosorbent Assay
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Applications Microscopy
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 26-2F recognizes human angiogenin. Angiogenin is a secreted ribonuclease (RNASE)-family member and is important for the formation of new blood vessels as well as the stimulation of cell proliferation and survival. Cellular stress signals direct angiogenin to stress granules where angiogenin functions by hydrolyzing cellular tRNA resulting in modulation of protein translation and promotion of cell survival. Cytoplasmic angiogenin can bind to actin, forming complexes that activate protease cascades which can degrade laminin and fibronectin allowing endothelial cells to penetrate the basement membrane and migrate. In cancer cells, growth signals direct angiogenin translocation to the nucleus where binding of DNA promotor regions stimulates rRNA transcription resulting in a downstream increase in protein synthesis driving cell growth and survival. Angiogenin is upregulated in multiple cancers including breast, prostate, cervical, colorectal, lymphoma, and melanoma. During development and in adulthood, angiogenin is highly expressed in the spinal cord and contributes to motor neuron survival. Angiogenin is important for neuronal survival and is depleted in neurodegenerative diseases such as amyotrophic lateral sclerosis (ALS) and Parkinson’s disease. This 26-2F antibody is reported to have both in vivo and in vitro neutralizing activity to angiogenin.