missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Angiogenin (108-123)

For cellular and molecular biology applications

Specifications

Product Type Angiogenin (108-123)
Molecular Weight 1878.09
Storage Requirements -30° to -10°C
Sequence Glu-Asn-Gly-Leu-Pro-Val-His-Leu-Asp-Gln-Ser-Ile-Phe-Arg-Arg-Pro
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66320LB0
View Documents
Alfa Aesar™
J66320LB0
5mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ66320MA
View Documents
Alfa Aesar™
J66320MA
10mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

Angiogenin (108-123)
-30° to -10°C
Solid
HPLC
1878.09
Glu-Asn-Gly-Leu-Pro-Val-His-Leu-Asp-Gln-Ser-Ile-Phe-Arg-Arg-Pro
RUO

For Research Use Only