Please login to your online account to display your discounted pricing

Alfa Aesar™ Angiogenin (108-122)

For cellular and molecular biology applications

Specifications

Product Type Angiogenin (108-122)
Molecular Weight 1780.98
Storage Requirements -30° to -10°C
Sequence Glu-Asn-Gly-Leu-Pro-Val-His-Leu-Asp-Gln-Ser-Ile-Phe-Arg-Arg
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66187LB0

 
alfa aesar™
J66187LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66187MA

 
alfa aesar™
J66187MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Angiogenin (108-122)
Molecular Weight 1780.98
Storage Requirements -30° to -10°C
Sequence Glu-Asn-Gly-Leu-Pro-Val-His-Leu-Asp-Gln-Ser-Ile-Phe-Arg-Arg
Format Solid
Regulatory Status RUO
Purification Method HPLC