missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anganese(II) sulfate hydrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1080706

 View more versions of this product

Catalog No. AA1080706



Specifications

Specifications

Manganese(II) sulfate hydrate
15244-36-7
H2MnO5S
Pink Crystalline Powder
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese(2+);sulfate;hydrate
177577
151.00 (Anhydrous)
Odorless
5g
Loses all water at 400-450°
99.999% (Metal basis)
MFCD00149159
manganese 2+ sulfate hydrate, manganese 2+ sulfate monohydrate, manganese ii sulfate hydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese sulfate monohydrate, manganese sulfate usp, manganous sulfate monohydrate, mnso4.h2o, unii-w00lys4t26
O.[O-]S(=O)(=O)[O-].[Mn+2]
169.009
CHEBI:86364
Crystalline Powder
Pink

Chemical Identifiers

15244-36-7
169.009
ISPYRSDWRDQNSW-UHFFFAOYSA-L
177577
manganese(2+);sulfate;hydrate
H2MnO5S
MFCD00149159
manganese 2+ sulfate hydrate, manganese 2+ sulfate monohydrate, manganese ii sulfate hydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese sulfate monohydrate, manganese sulfate usp, manganous sulfate monohydrate, mnso4.h2o, unii-w00lys4t26
CHEBI:86364
O.[O-]S(=O)(=O)[O-].[Mn+2]
Safety and Handling

Safety and Handling

H373-H411-H401: May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 232-089-9

Documents