Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Androgen Receptor Ab-2, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Androgen Receptor Ab-2, Rabbit Polyclonal Antibody.

$271.27 - $1,380.31

Specifications

Antigen Androgen Receptor Ab-2
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1358P

 
thermo scientific™ lab vision™
RB1358P
Purified Ab with BSA and Azide 1mL Each for $1,256.61

RB1358P0

 
thermo scientific™ lab vision™
RB1358P0
Purified Ab with BSA and Azide 100μL Each for $271.27

RB1358P1

 
thermo scientific™ lab vision™
RB1358P1
Purified Ab with BSA and Azide 500μL Each for $760.47

RB1358P1ABX

 
thermo scientific™ lab vision™
RB1358P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $844.49

RB1358PABX

 
thermo scientific™ lab vision™
RB1358PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,380.31
Description & Specifications

Specifications

Antigen Androgen Receptor Ab-2
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Immunogen A synthetic peptide derived from N-terminus of rat AR
Isotype IgG

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Androgen Receptor Ab-2, Rabbit Polyclonal Antibody.

The expression of androgen receptor (AR) is reportedly inversely correlated with histologic grade, i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed as a marker of hormone-responsiveness.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide derived from N-terminus of rat AR

Molecular Weight: 110kDa

Positive Control: LnCAp cells or prostate CA

Cellular Localization: Nuclear

Ab-2 is suitable for staining of formalin/paraffin tissues.

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO