Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Androgen Receptor Ab-2, Rabbit Polyclonal Antibody


Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Androgen Receptor Ab-2, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1358PABX

 View more versions of this product

Catalog No. RB1358PABX

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Antigen Androgen Receptor Ab-2
Immunogen A synthetic peptide derived from N-terminus of rat AR
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG
Quantity 1mL
Host Species Rabbit
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Oncogenesis
Classification Polyclonal
Conjugate Unlabeled
Type Primary
Concentration 1mg/mL
Item Description 1mL; 1mg/mL; Unlabeled

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Androgen Receptor Ab-2, Rabbit Polyclonal Antibody.

The expression of androgen receptor (AR) is reportedly inversely correlated with histologic grade, i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed as a marker of hormone-responsiveness.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide derived from N-terminus of rat AR

Molecular Weight: 110kDa

Positive Control: LnCAp cells or prostate CA

Cellular Localization: Nuclear

Ab-2 is suitable for staining of formalin/paraffin tissues.

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO