Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Androgen Receptor Ab-2, Rabbit Polyclonal Antibody


Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Androgen Receptor Ab-2, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1358P1ABX

 View more versions of this product

Catalog No. RB1358P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Androgen Receptor Ab-2
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Rabbit
Immunogen A synthetic peptide derived from N-terminus of rat AR
Isotype IgG
Quantity 500μL
Regulatory Status RUO
Research Discipline Oncogenesis
Type Primary
Monoclonal or Polyclonal Polyclonal
Target Species Human
Target Species Murine

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Androgen Receptor Ab-2, Rabbit Polyclonal Antibody.

The expression of androgen receptor (AR) is reportedly inversely correlated with histologic grade, i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed as a marker of hormone-responsiveness.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide derived from N-terminus of rat AR

Molecular Weight: 110kDa

Positive Control: LnCAp cells or prostate CA

Cellular Localization: Nuclear

Ab-2 is suitable for staining of formalin/paraffin tissues.

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO