Please login to your online account to display your discounted pricing

Anatomy Dissection Mats


Resistant to spills and easily cleaned with a sponge or paper towel.

Manufacturer: fisher science education  420.5060

 View more versions of this product

Catalog No. S66915

  • / Each
 

Description & Specifications

Specifications

Includes One each of cat, crayfish, earthworm, fetal pig, frog and grasshopper
Product Type Dissection anatomy mat
Category Biology
For Use With (Application) Resistant to spills and easily cleaned with a sponge or paper towel
Format Set