Please login to your online account to display your discounted pricing

Anastrozole, Tocris

Potent aromatase (CYP19) inhibitor

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material Anastrozole
Assay Percent Range >99%
Molecular Formula C17H19N5
Formula Weight 293.37
Synonym α1,α1,α3,α3-Tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

338810

 
r & d systems
338810
10mg Each for $239.63

338850

 
r & d systems
338850
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 293.37
Quantity 10mg
CAS 120511-73-1
Assay Percent Range >99%
Chemical Name or Material Anastrozole
Molecular Formula C17H19N5
Synonym α1,α1,α3,α3-Tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature