Ohaus™ Analog Dry Block Heater
OHAUS Analog Dry Block Heaters are ideal for applications that require temperature stability. The close tube-to-block contact enables maximum heat retention, resulting in efficient heating
Manufacturer:  Ohaus™ 30392073
 View more versions of this product
Catalog No. S72094
Description
Description
  • High-wattage, constant temp. analog models are an economical option
  • Multi-purpose use
  • Holds interchangeable modular blocks (sold separately)
  • Analog controls
Specifications
Specifications
Analog
Ohaus Modular Blocks
3.5 in.
3 wire cord and plug
53.1 cm
±0.1°C
6 Block
410 w
8 in.
5°C to 100°C low range; 75°C to 150°C high range
50/60 Hz
8.9 cm
20.9 in.
±2°C at 37°C
No
120 V
4.5 kg
20.3 cm
Documents