Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-Anabasine, tech. 85%

Chemical Identifiers

Chemical Name or Material (+/-)-Anabasine
CAS 13078-04-1
Purity Grade Technical Grade
Molecular Formula C10H14N2
Formula Weight 162.24
MDL Number MFCD00006370
UN Number UN3140
Beilstein 82639
Merck Index 14,619
Synonym Neonicotine; 2-(3-Pyridyl)piperidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1208903

  SDS
alfa aesar™
L1208903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.24
Chemical Name or Material (+/-)-Anabasine
MDL Number MFCD00006370
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H227
Fatal if swallowed.
Fatal in contact with skin.
Combustible liquid.
Beilstein 82639
Assay 85%
Merck Index 14,619
UN Number UN3140
Health Hazard 3 GHS P Statement
P210-P301+P310-P361-P302+P350-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 93°C (199°F)
Quantity 1g
RTECS Number BV4375000
Boiling Point 270° to 272°C
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALKALOIDS, LIQUID, N.O.S.
Purity Grade Technical Grade
Melting Point ≈9°C
CAS 13078-04-1
Molecular Formula C10H14N2
Synonym Neonicotine; 2-(3-Pyridyl)piperidine
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P361-P302+P350-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H227
Fatal if swallowed.
Fatal in contact with skin.
Combustible liquid.