missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-Anabasine, tech. 85%

Manufacturer:  Alfa Aesar™ L1208903

 View more versions of this product

Catalog No. AAL1208903



Specifications

Specifications

(+/-)-Anabasine
270°C to 272°C
1g
C10H14N2
UN3140
14,619
MTXSIJUGVMTTMU-UHFFFAOYSA-N
3-piperidin-2-ylpyridine
2181
162.24
Technical
93°C (199°F)
~9°C
13078-04-1
MFCD00006370
82639
3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl
C1CCNC(C1)C2=CN=CC=C2
162.236
CHEBI:28986
85%

Chemical Identifiers

13078-04-1
162.236
MTXSIJUGVMTTMU-UHFFFAOYSA-N
2181
3-piperidin-2-ylpyridine
C10H14N2
MFCD00006370
3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl
CHEBI:28986
C1CCNC(C1)C2=CN=CC=C2
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H227
Fatal if swallowed.
Fatal in contact with skin.
Combustible liquid.

GHS P Statement
P210-P301+P310-P361-P302+P350-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALKALOIDS, LIQUID, N.O.S.

RTECSNumber : BV4375000

Documents