Please login to your online account to display your discounted pricing

Alfa Aesar™ Amyloid beta (35-25)

For cellular and molecular biology applications

Specifications

Product Type Amyloid beta (35-25)
Molecular Weight 1060.27
Storage Requirements -30° to -10°C
Format Solid
Sequence Met-Leu-Gly-Ile-Ile-Ala-Gly-Lys-Asn-Ser-Gly
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66826EXD

 
alfa aesar™
J66826EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66826MCR

 
alfa aesar™
J66826MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Amyloid beta (35-25)
Molecular Weight 1060.27
Storage Requirements -30° to -10°C
Format Solid
Sequence Met-Leu-Gly-Ile-Ile-Ala-Gly-Lys-Asn-Ser-Gly
Regulatory Status RUO
Purification Method HPLC