Please login to your online account to display your discounted pricing

Alfa Aesar™ Amyloid beta (25-35)

For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66808-EXD

 View all options for this product

Catalog No. AAJ66808EXD

 Please call Customer Service at 1-800-234-7437 or send an email to for assistance.

Description & Specifications


Molecular Weight 1060.27
Purification Method HPLC
Quantity 0.5mg
Format Solid
Sequence Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Amyloid beta (25-35)