Please login to your online account to display your discounted pricing

Alfa Aesar™ Amyloid beta (25-35)

For cellular and molecular biology applications

Specifications

Product Type Amyloid beta (25-35)
Molecular Weight 1060.27
Storage Requirements -30° to -10°C
Format Solid
Sequence Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66808MCR

 
alfa aesar™
J66808MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Amyloid beta (25-35)
Molecular Weight 1060.27
Storage Requirements -30° to -10°C
Format Solid
Sequence Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met
Regulatory Status RUO
Purification Method HPLC