Please login to your online account to display your discounted pricing

Alfa Aesar™ Amyloid beta (22-35)

For cellular and molecular biology applications

Specifications

Product Type Amyloid beta (22-35)
Molecular Weight 1403.60
Storage Requirements -30° to -10°C
Sequence Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66715EXD

 
alfa aesar™
J66715EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66715MCR

 
alfa aesar™
J66715MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Amyloid beta (22-35)
Molecular Weight 1403.60
Storage Requirements -30° to -10°C
Sequence Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met
Format Solid
Regulatory Status RUO
Purification Method HPLC