Please login to your online account to display your discounted pricing

Alfa Aesar™ Amyloid beta (22-35)


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66715MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66715MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Quantity 1mg
Format Solid
Regulatory Status RUO
Sequence Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met
Product Type Amyloid beta (22-35)
Molecular Weight 1403.60
Purification Method HPLC