Please login to your online account to display your discounted pricing

Alfa Aesar™ Amyloid beta (1-38)

For cellular and molecular biology applications

Specifications

Product Type Amyloid beta (1-38)
Molecular Weight 4131.53
Storage Requirements -30° to -10°C
Format Solid
Sequence Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66913EXD

 
alfa aesar™
J66913EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66913MCR

 
alfa aesar™
J66913MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Amyloid beta (1-38)
Molecular Weight 4131.53
Storage Requirements -30° to -10°C
Format Solid
Sequence Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-
Regulatory Status RUO
Purification Method HPLC