Please login to your online account to display your discounted pricing

Alfa Aesar™ Amyloid beta (1-28)

For cellular and molecular biology applications

Specifications

Product Type Amyloid beta (1-28)
Molecular Weight 3262.45
Storage Requirements -30° to -10°C
Sequence Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66757EXD

 
alfa aesar™
J66757EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66757MCR

 
alfa aesar™
J66757MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Amyloid beta (1-28)
Molecular Weight 3262.45
Storage Requirements -30° to -10°C
Sequence Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys
Format Solid
Regulatory Status RUO
Purification Method HPLC