missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Amyloid A4/Amyloid β Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Amyloid A4/Amyloid β Ab-1, Mouse Monoclonal Antibody.

$283.04 - $1,191.63

Specifications

Antigen Amyloid A4/Amyloid beta Ab-1
Clone BAM01; same as 6F/3D
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MS1167R7 Thermo Scientific™ Lab Vision™
MS1167R7
7mL Each for $685.51
MS1167S Thermo Scientific™ Lab Vision™
MS1167S
1mL Each for $1,191.63
MS1167S0 Thermo Scientific™ Lab Vision™
MS1167S0
100μL Each for $283.04
MS1167S1 Thermo Scientific™ Lab Vision™
MS1167S1
500μL Each for $794.20
Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Amyloid A4/Amyloid β Ab-1, Mouse Monoclonal Antibody.

Amyloid beta-protein/amyloid A4 or amyloid beta-peptide is derived from the amyloid beta-precursor protein. Beta amyloid is an extracellular filamentous protein deposit found in the brain. It is the major protein component of amyloid cores, neuritic plaques and is also found as a deposit in neurofibrillary tangles. In humans, Alzheimer's disease is the most common cause of senile dementia and is characterized by abnormal filamentous protein deposits in the brain. Beta amyloid deposits are also detected in Lewy body dementia, Down's syndrome, amyloidosis (Dutch type) and in the Guam Parkinson-Dementia complex.

Host Species: Mouse

Clone: BAM01; same as 6F/3D

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: aa 8-17 (SGYEVHHQKL) of mature amyloid A4/ amyloid Beta which is the same as aa 660-669 of human amyloid A4/amyloid β protein precursor

Immunogen: A synthetic peptide consisting of residues 8-17 (SGYEVHHQKL) of the amyloid β-protein/amyloid A4 which is the same as aa 660-669 of human amyloid A4/amyloid β protein precursor

Molecular Weight: 102kDa (precursor), 4-5kDa (mature)

Positive Control: Human brain of Alzheimer's disease

Cellular Localization: Transmembrane (APP) and extracellular (Amyloid β)

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

.
Description & Specifications

Specifications

Amyloid A4/Amyloid beta Ab-1
Immunohistochemistry (Paraffin)
Unlabeled
RUO
Primary
IgG2a kappa
BAM01; same as 6F/3D
Monoclonal
Murine
Neuroscience
A synthetic peptide consisting of residues 8-17 (SGYEVHHQKL) of the amyloid beta-protein/amyloid A4 which is the same as aa 660-669 of human amyloid A4 / amyloid Beta protein precursor